Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)benzylamine, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  312950100

 View more versions of this product

Catalog No. AC312950100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 175.16
Quantity 10g
Color Clear colorless to light yellow
EINECS Number 221-258-2
TSCA TSCA
Density 1.249g/mL
CAS 419308
CAS Max % 100.0
Chemical Name or Material 2-(Trifluoromethyl)benzylamine
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H8F3N
Linear Formula CF3C6H4CHNH
MDL Number MFCD00010297
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4700 to 1.4720 (20°C, 589nm)

Specifications

Chemical Name or Material 2-(Trifluoromethyl)benzylamine
Name Note 98%
CAS 419308
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8F3N
Linear Formula CF3C6H4CHNH
Formula Weight 175.16
MDL Number MFCD00010297
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents