Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzyl chloride
CAS 21742-00-7
Molecular Formula C8H6ClF3
Formula Weight 194.59
MDL Number MFCD00009920
UN Number UN3265
Beilstein 389380
Synonym alpha'-Chloro-alpha,alpha,alpha-trifluoro-o-xylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2064806

  SDS
alfa aesar™
B2064806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2064814

  SDS
alfa aesar™
B2064814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.59
Chemical Name or Material 2-(Trifluoromethyl)benzyl chloride
Odor Pungent
Assay ≥98%
Molecular Formula C8H6ClF3
MDL Number MFCD00009920
CAS 21742-00-7
UN Number UN3265
Beilstein 389380
Synonym alpha'-Chloro-alpha,alpha,alpha-trifluoro-o-xylene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 70°C (158°F)
Boiling Point 67° to 68°C (12mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.