missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)benzyl Bromide, TCI America™

Manufacturer:  TCI America T276825G

 View more versions of this product

Catalog No. T276825G



Specifications

Specifications

2-(Trifluoromethyl)benzyl Bromide
White-Yellow
395-44-8
MFCD00013558
2-trifluoromethyl benzyl bromide, 1-bromomethyl-2-trifluoromethyl benzene, 2-trifluoromethylbenzyl bromide, 2-trifluoromethyl benzylbromide, o-trifluoromethylbenzyl bromide, benzene, 1-bromomethyl-2-trifluoromethyl, o-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-o-xylene, 1-bromomethyl-2-trifluoromethyl-benzene, a'-bromo-a,a,a-trifluoro-o-xylene
C1=CC=C(C(=C1)CBr)C(F)(F)F
239.035
239.04
≥97.0% (GC,T)
72°C
25g
C8H6BrF3
3261
TXVVVEUSVBLDED-UHFFFAOYSA-N
1-(bromomethyl)-2-(trifluoromethyl)benzene
123057
Crystal-Lump at 20°C

Chemical Identifiers

395-44-8
239.035
TXVVVEUSVBLDED-UHFFFAOYSA-N
123057
C1=CC=C(C(=C1)CBr)C(F)(F)F
C8H6BrF3
MFCD00013558
2-trifluoromethyl benzyl bromide, 1-bromomethyl-2-trifluoromethyl benzene, 2-trifluoromethylbenzyl bromide, 2-trifluoromethyl benzylbromide, o-trifluoromethylbenzyl bromide, benzene, 1-bromomethyl-2-trifluoromethyl, o-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-o-xylene, 1-bromomethyl-2-trifluoromethyl-benzene, a'-bromo-a,a,a-trifluoro-o-xylene
1-(bromomethyl)-2-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Documents