Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzyl bromide, 97+%

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzyl bromide
CAS 395-44-8
Molecular Formula C8H6BrF3
Formula Weight 239.04
MDL Number MFCD00013558
UN Number UN3261
Beilstein 2256977
Synonym alpha'-Bromo-alpha,alpha,alpha-trifluoro-o-xylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2063403

  SDS
alfa aesar™
B2063403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2063406

  SDS
alfa aesar™
B2063406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2063414

  SDS
alfa aesar™
B2063414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.04
Chemical Name or Material 2-(Trifluoromethyl)benzyl bromide
Odor Pungent
Assay ≥97%
Molecular Formula C8H6BrF3
MDL Number MFCD00013558
CAS 395-44-8
UN Number UN3261
Beilstein 2256977
Synonym alpha'-Bromo-alpha,alpha,alpha-trifluoro-o-xylene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Flash Point 83°C (181°F)
Boiling Point 71° to 72°C (7.5mmHg)
Melting Point 27° to 30°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.