Learn More
Alfa Aesar™ 2-(Trifluoromethyl)benzyl alcohol, 97%
Manufacturer: Alfa Aesar™ A1566714
Specifications
2-(Trifluoromethyl)benzyl alcohol | |
80°C to 83°C (3mmHg) | |
25g | |
C8H7F3O | |
2093714 | |
TWQNSHZTQSLJEE-UHFFFAOYSA-N | |
[2-(trifluoromethyl)phenyl]methanol | |
67666 | |
97% |
92°C (197°F) | |
4°C | |
346-06-5 | |
MFCD00004621 | |
2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol | |
C1=CC=C(C(=C1)CO)C(F)(F)F | |
176.138 | |
176.14 |
Chemical Identifiers
346-06-5 | |
176.138 | |
TWQNSHZTQSLJEE-UHFFFAOYSA-N | |
67666 | |
C1=CC=C(C(=C1)CO)C(F)(F)F |
C8H7F3O | |
MFCD00004621 | |
2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol | |
[2-(trifluoromethyl)phenyl]methanol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning