missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzyl alcohol, 97%

Manufacturer:  Alfa Aesar™ A1566703

 View more versions of this product

Catalog No. AAA1566703



Specifications

Specifications

2-(Trifluoromethyl)benzyl alcohol
80°C to 83°C (3mmHg)
1g
C8H7F3O
2093714
TWQNSHZTQSLJEE-UHFFFAOYSA-N
[2-(trifluoromethyl)phenyl]methanol
67666
97%
92°C (197°F)
4°C
346-06-5
MFCD00004621
2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol
C1=CC=C(C(=C1)CO)C(F)(F)F
176.138
176.14

Chemical Identifiers

346-06-5
176.138
TWQNSHZTQSLJEE-UHFFFAOYSA-N
67666
C1=CC=C(C(=C1)CO)C(F)(F)F
C8H7F3O
MFCD00004621
2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol
[2-(trifluoromethyl)phenyl]methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents