Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzyl alcohol, 97%

$18.32 - $18.32

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzyl alcohol
CAS 346-06-5
Molecular Formula C8H7F3O
Formula Weight 176.14
MDL Number MFCD00004621
Beilstein 2093714
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1566703

 
alfa aesar™
A1566703
1g Each for $18.32
Description & Specifications

Specifications

Formula Weight 176.14
Chemical Name or Material 2-(Trifluoromethyl)benzyl alcohol
Assay 97%
Molecular Formula C8H7F3O
MDL Number MFCD00004621
CAS 346-06-5
Beilstein 2093714
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 92°C (197°F)
Boiling Point 80° to 83°C (3mmHg)
Melting Point 4°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.