Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2- (Trifluoromethyl)benzoyl chloride
CAS 312-94-7
Molecular Formula C8H4ClF3O
Formula Weight 208.57
MDL Number MFCD00000667
UN Number UN3265
Beilstein 881040
Synonym alpha,alpha,alpha-Trifluoro-o-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1056406

 
alfa aesar™
A1056406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1056414

 
alfa aesar™
A1056414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1056422

 
alfa aesar™
A1056422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.57
Chemical Name or Material 2- (Trifluoromethyl)benzoyl chloride
Odor Pungent
Assay 98%
Molecular Formula C8H4ClF3O
MDL Number MFCD00000667
CAS 312-94-7
UN Number UN3265
Beilstein 881040
Synonym alpha,alpha,alpha-Trifluoro-o-toluoyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 95°C (203°F)
Boiling Point 89° to 90°C (16mmHg)
Melting Point −22°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.