Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzophenone, 98%

$92.66 - $394.05

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzophenone
CAS 727-99-1
Molecular Formula C14H9F3O
Formula Weight 250.22
MDL Number MFCD00000377
Beilstein 2375025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2022106

 
alfa aesar™
B2022106
5g Each for $92.66

AAB2022114

 
alfa aesar™
B2022114
25g Each for $394.05
Description & Specifications

Specifications

Formula Weight 250.22
Chemical Name or Material 2-(Trifluoromethyl)benzophenone
Assay 98%
Molecular Formula C14H9F3O
MDL Number MFCD00000377
CAS 727-99-1
Beilstein 2375025
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 58° to 61°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.