missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ A1495118

 View more versions of this product

Catalog No. AAA1495118



Specifications

Specifications

2-(Trifluoromethyl)benzonitrile
204°C to 206°C
50g
C8H4F3N
UN3276
2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
C1=CC=C(C(=C1)C#N)C(F)(F)F
171.122
171.12
90°C (194°F)
17°C to 18°C
447-60-9
MFCD00001791
2048152
SOZGHDCEWOLLHV-UHFFFAOYSA-N
2-(trifluoromethyl)benzonitrile
67972
97%

Chemical Identifiers

447-60-9
171.122
SOZGHDCEWOLLHV-UHFFFAOYSA-N
67972
C1=CC=C(C(=C1)C#N)C(F)(F)F
C8H4F3N
MFCD00001791
2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
2-(trifluoromethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Documents