Learn More
2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
$74.66 - $74.66
Chemical Identifiers
| CAS | 433-97-6 |
|---|---|
| Molecular Formula | C8H5F3O2 |
| Molecular Weight (g/mol) | 190.12 |
| MDL Number | MFCD00002476 |
| InChI Key | FBRJYBGLCHWYOE-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid |
| PubChem CID | 9899 |
| ChEBI | CHEBI:60694 |
| IUPAC Name | 2-(trifluoromethyl)benzoic acid |
| SMILES | OC(=O)C1=CC=CC=C1C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC188450050
|
Thermo Scientific Chemicals
188450050 |
5 g | Glass bottle |
Each for $74.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 433-97-6 | |
| 190.12 | |
| FBRJYBGLCHWYOE-UHFFFAOYSA-N | |
| 9899 | |
| 2-(trifluoromethyl)benzoic acid |
| C8H5F3O2 | |
| MFCD00002476 | |
| 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid | |
| CHEBI:60694 | |
| OC(=O)C1=CC=CC=C1C(F)(F)F |
Specifications
| 433-97-6 | |
| 100.0 | |
| Brown-Yellow to Yellow | |
| Authentic | |
| Glass bottle | |
| CF3C6H4CO2H | |
| 5 g | |
| 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid | |
| OC(=O)C1=CC=CC=C1C(F)(F)F | |
| 190.12 | |
| CHEBI:60694 | |
| 98% | |
| 2-(Trifluoromethyl)benzoic acid, 98% |
| 97.5 | |
| 107.0°C to 110.0°C | |
| 247.0°C (753.0 mmHg) | |
| 97.5% min. (HPLC) | |
| C8H5F3O2 | |
| MFCD00002476 | |
| 09, III, 2308 | |
| FBRJYBGLCHWYOE-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)benzoic acid | |
| 9899 | |
| 190.12 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 207-093-9
TSCA : TSCA
RUO – Research Use Only