Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzoic acid, 98+%

$94.31 - $1,116.51

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzoic acid
CAS 433-97-6
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD00002476
Beilstein 976984
Synonym alpha,alpha,alpha-Trifluoro-o-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1609306

  SDS
alfa aesar™
A1609306
5g Each for $94.31

AAA1609314

  SDS
alfa aesar™
A1609314
25g Each for $319.43

AAA1609322

  SDS
alfa aesar™
A1609322
100g Each for $1,116.51
Description & Specifications

Specifications

Formula Weight 190.12
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H5F3O2
Boiling Point 247° to 254°C
Beilstein 976984
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Quantity 5g
Melting Point 107° to 110°C
Chemical Name or Material 2-(Trifluoromethyl)benzoic acid
CAS 433-97-6
Health Hazard 1 Warning
MDL Number MFCD00002476
Synonym alpha,alpha,alpha-Trifluoro-o-toluic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.