Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzenesulfonamide, 97%

$94.31 - $337.51

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzenesulfonamide
CAS 1869-24-5
Molecular Formula C7H6F3NO2S
Formula Weight 225.19
MDL Number MFCD00042420
Beilstein 2697078
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1971703

  SDS
alfa aesar™
L1971703
1g Each for $94.31

AAL1971706

  SDS
alfa aesar™
L1971706
5g Each for $337.51
Description & Specifications

Specifications

Formula Weight 225.19
Chemical Name or Material 2-(Trifluoromethyl)benzenesulfonamide
CAS 1869-24-5
Beilstein 2697078
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 97%
Molecular Formula C7H6F3NO2S
MDL Number MFCD00042420
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 188° to 192°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.