Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzeneboronic acid, 97%

$63.73 - $1,140.62

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzeneboronic acid
CAS 1423-27-4
Molecular Formula C7H6BF3O2
Formula Weight 189.93
MDL Number MFCD00236059
Beilstein 2974465
Synonym 2-(Trifluoromethyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2434303

  SDS
alfa aesar™
B2434303
1g Each for $63.73

AAB2434306

  SDS
alfa aesar™
B2434306
5g Each for $250.12

AAB2434314

  SDS
alfa aesar™
B2434314
25g Each for $1,140.62
Description & Specifications

Specifications

Formula Weight 189.93
Health Hazard 1 Warning
CAS 1423-27-4
Melting Point 108° to 110°C
MDL Number MFCD00236059
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-(Trifluoromethyl)benzeneboronic acid
Assay 97%
Molecular Formula C7H6BF3O2
Synonym 2-(Trifluoromethyl)phenylboronic acid
Beilstein 2974465
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.