Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)benzamide, 98+%

$345.05 - $345.05

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzamide
CAS 360-64-5
Molecular Formula C8H6F3NO
Formula Weight 189.14
MDL Number MFCD00014798
Beilstein 2616148
Synonym alpha,alpha,alpha-Trifluoro-o-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1410314

 
alfa aesar™
A1410314
25g Each for $345.05
Description & Specifications

Specifications

Formula Weight 189.14
Chemical Name or Material 2-(Trifluoromethyl)benzamide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00014798
Beilstein 2616148
Melting Point 161° to 164°C
Synonym alpha,alpha,alpha-Trifluoro-o-toluamide
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H6F3NO
Health Hazard 1 Warning
Assay ≥98%
CAS 360-64-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.