missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethyl)benzaldehyde, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC232681000
Specifications
2-(Trifluoromethyl)benzaldehyde | |
61°C | |
Glass bottle | |
1.32 | |
Colorless to Yellow | |
100g | |
96.0 | |
96% min. (GC) | |
CF3C6H4CHO | |
07, III, 1013 | |
ZDVRPKUWYQVVDX-UHFFFAOYSA-N | |
2-(trifluoromethyl)benzaldehyde | |
123067 | |
Liquid |
1.3200g/mL | |
Authentic | |
1.4650 to 1.4670 | |
70.0°C to 71.0°C (16.0mmHg) | |
-40.0°C | |
447-61-0 | |
100.0 | |
C8H5F3O | |
MFCD00003337 | |
2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-benzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde | |
C1=CC=C(C(=C1)C=O)C(F)(F)F | |
174.12 | |
174.12 | |
97% |
Chemical Identifiers
447-61-0 | |
174.12 | |
ZDVRPKUWYQVVDX-UHFFFAOYSA-N | |
123067 | |
C1=CC=C(C(=C1)C=O)C(F)(F)F |
C8H5F3O | |
MFCD00003337 | |
2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-benzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde | |
2-(trifluoromethyl)benzaldehyde |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 207-185-9