Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)benzaldehyde, 97%, ACROS Organics™

$91.22 - $91.22

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzaldehyde
CAS 447-61-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5F3O
Linear Formula CF3C6H4CHO
Formula Weight 174.12
MDL Number MFCD00003337
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC232680250

  SDS
thermo fisher scientific
AC232680250
Glass bottle 25g Each for $91.22

AC232681000

  SDS
thermo fisher scientific
AC232681000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 174.12
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 07, III, 1013
Density 1.32g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-(Trifluoromethyl)benzaldehyde
Assay Percent Range 97%
Molecular Formula C8H5F3O
Linear Formula CF3C6H4CHO
MDL Number MFCD00003337
CAS 447-61-0
Infrared Spectrum Authentic
Synonym α, α, α-Trifluoro-o-tolualdehyde
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.