Learn More
2-(Trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1507222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(Trifluoromethyl)aniline | |
-35°C to -33°C | |
67°C to 68°C (15 mmHg) | |
Recognizable | |
1.48 | |
100 g | |
879494 | |
VBLXCTYLWZJBKA-UHFFFAOYSA-N | |
2-(trifluoromethyl)aniline | |
6922 | |
98% |
88-17-5 | |
1.295 | |
65°C (149°F) | |
C7H6F3N | |
MFCD00007718 | |
UN2942 | |
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene | |
C1=CC=C(C(=C1)C(F)(F)F)N | |
161.127 | |
161.13 |
Chemical Identifiers
88-17-5 | |
161.127 | |
VBLXCTYLWZJBKA-UHFFFAOYSA-N | |
6922 | |
C1=CC=C(C(=C1)C(F)(F)F)N |
C7H6F3N | |
MFCD00007718 | |
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene | |
2-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H318-H373-H302-H335-H227-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
P210-P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P314-P330-P333+P313-P362-P370+P378q-P501c
H227-H302-H315-H317-H319-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-TRIFLUOROMETHYLANILINE
EINECSNumber : 201-806-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only