missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)aniline, 98%

Manufacturer:  Alfa Aesar™ A1507236

 View more versions of this product

Catalog No. AAA1507236



Specifications

Specifications

2-(Trifluoromethyl)aniline
Recognizable
-35°C to -33°C
88-17-5
MFCD00007718
879494
VBLXCTYLWZJBKA-UHFFFAOYSA-N
2-(trifluoromethyl)aniline
6922
98%
65°C (149°F)
67°C to 68°C (15mmHg)
500g
C7H6F3N
UN2942
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
C1=CC=C(C(=C1)C(F)(F)F)N
161.127
161.13

Chemical Identifiers

88-17-5
161.127
VBLXCTYLWZJBKA-UHFFFAOYSA-N
6922
C1=CC=C(C(=C1)C(F)(F)F)N
C7H6F3N
MFCD00007718
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
2-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H318-H373-H302-H335-H227-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

GHS P Statement
P280-P273-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-TRIFLUOROMETHYLANILINE

TSCA : TSCA

Documents