Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)aniline
CAS 88-17-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H6F3N
Linear Formula H2NC6H4CF3
Formula Weight 161.13
MDL Number MFCD00007718
Synonym 2-(Trifluoromethyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00885EA

  SDS
fisher scientific company
SB00885EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.13
Quantity 10g
Assay 97%
Density 1.2960g/mL
CAS Min % 97.0
Linear Formula H2NC6H4CF3
Synonym 2-(Trifluoromethyl)aniline
CAS 88-17-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Molecular Formula C7H6F3N
MDL Number MFCD00007718
Chemical Name or Material 2-(Trifluoromethyl)aniline
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%