Please login to your online account to display your discounted pricing

2'-(Trifluoromethyl)acetophenone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2';-(Trifluoromethyl)acetophenone
CAS 17408-14-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
Formula Weight 188.15
MDL Number MFCD00000378
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC214430010

  SDS
thermo fisher scientific
214430010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 188.15
Quantity 1g
Color Clear colorless to very slightly yellow
Density 1.2500g/mL
CAS Max % 100.0
Chemical Name or Material 2';-(Trifluoromethyl)acetophenone
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
CAS 17408-14-9
Infrared Spectrum Authentic
Refractive Index 1.4584 to 1.4604 (20°C, 589nm)
MDL Number MFCD00000378
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.