Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-(Trifluoromethyl)acetophenone, 97+%

$346.55 - $346.55

Chemical Identifiers

Chemical Name or Material 2'- (Trifluoromethyl)acetophenone
CAS 17408-14-9
Molecular Formula C9H7F3O
Formula Weight 188.15
MDL Number MFCD00000378
Beilstein 2259035
Synonym 2-Acetylbenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1224814

  SDS
alfa aesar™
A1224814
25g Each for $346.55
Description & Specifications

Specifications

Formula Weight 188.15
Chemical Name or Material 2'- (Trifluoromethyl)acetophenone
Odor Characteristic
Assay ≥97%
Molecular Formula C9H7F3O
MDL Number MFCD00000378
CAS 17408-14-9
Beilstein 2259035
Synonym 2-Acetylbenzotrifluoride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Flash Point 84°C (183°F)
Boiling Point 163° to 165°C
Melting Point 15° to 17°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.