Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-(Trifluoromethyl)acetanilide, 98%

$73.08 - $857.35

Chemical Identifiers

Chemical Name or Material 2'-(Trifluoromethyl)acetanilide
CAS 344-62-7
Molecular Formula C9H8F3NO
Formula Weight 203.16
MDL Number MFCD00013555
Beilstein 2106815
Synonym 2-Acetamidobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1659014

 
alfa aesar™
A1659014
25g Each for $272.73

AAA1659006

 
alfa aesar™
A1659006
5g Each for $73.08

AAA1659022

 
alfa aesar™
A1659022
100g Each for $857.35
Description & Specifications

Specifications

Formula Weight 203.16
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
MDL Number MFCD00013555
Beilstein 2106815
Molecular Formula C9H8F3NO
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number AN3760000
Assay 98%
Chemical Name or Material 2'-(Trifluoromethyl)acetanilide
Odor Odorless
Melting Point 94° to 97°C
Quantity 25g
Synonym 2-Acetamidobenzotrifluoride
CAS 344-62-7
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.