Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)-6-pyridinecarboxylic acid
Assay Percent Range 97%
Molecular Formula C7H4F3NO2
Formula Weight 191.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC446750010

 
thermo fisher scientific
AC446750010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC446750050

 
thermo fisher scientific
AC446750050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-(Trifluoromethyl)-6-pyridinecarboxylic acid
Quantity 1g
CAS 131747-42-7
Assay Percent Range 97%
Molecular Formula C7H4F3NO2
Formula Weight 191.11