Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC446750050

 View more versions of this product

Catalog No. AC446750050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.11
Quantity 5g
Chemical Name or Material 2-(Trifluoromethyl)-6-pyridinecarboxylic acid
Assay Percent Range 97%
Molecular Formula C7H4F3NO2
CAS 131747-42-7

Specifications

Chemical Name or Material 2-(Trifluoromethyl)-6-pyridinecarboxylic acid
Assay Percent Range 97%
Molecular Formula C7H4F3NO2
Formula Weight 191.11