missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethyl)-1H-indole, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 51310-54-4 |
---|---|
Molecular Formula | C9H6F3N |
Molecular Weight (g/mol) | 185.15 |
InChI Key | QFHVHZJGQWMBTE-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, 1h-indole, 2-trifluoromethyl, pubchem23796, 2-trifluoromethyl indole |
PubChem CID | 10932124 |
IUPAC Name | 2-(trifluoromethyl)-1H-indole |
SMILES | C1=CC=C2C(=C1)C=C(N2)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC446560010
|
Thermo Scientific Chemicals
446560010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
51310-54-4 | |
185.15 | |
2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, 1h-indole, 2-trifluoromethyl, pubchem23796, 2-trifluoromethyl indole | |
2-(trifluoromethyl)-1H-indole |
C9H6F3N | |
QFHVHZJGQWMBTE-UHFFFAOYSA-N | |
10932124 | |
C1=CC=C2C(=C1)C=C(N2)C(F)(F)F |
Specifications
51310-54-4 | |
Glass Bottle | |
1g | |
QFHVHZJGQWMBTE-UHFFFAOYSA-N | |
2-(trifluoromethyl)-1H-indole | |
10932124 | |
97% |
96% min. (GC) | |
C9H6F3N | |
2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, 1h-indole, 2-trifluoromethyl, pubchem23796, 2-trifluoromethyl indole | |
C1=CC=C2C(=C1)C=C(N2)C(F)(F)F | |
185.15 | |
185.15 | |
2-(Trifluoromethyl)-1H-indole |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only