missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethyl)-1H-indole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446560050
Specifications
2-(Trifluoromethyl)-1H-indole | |
5g | |
96% min. (GC) | |
1h-indole, 2-trifluoromethyl, 2-trifluoromethyl indole, 2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, pubchem23796 | |
C1=CC=C2C(=C1)C=C(N2)C(F)(F)F | |
185.15 | |
185.15 |
107°C to 109°C | |
51310-54-4 | |
C9H6F3N | |
QFHVHZJGQWMBTE-UHFFFAOYSA-N | |
2-(trifluoromethyl)-1H-indole | |
10932124 | |
97% |
Chemical Identifiers
51310-54-4 | |
185.15 | |
1h-indole, 2-trifluoromethyl, 2-trifluoromethyl indole, 2-trifluoromethyl-1h-indole, 2-trifluoromethylindole, pubchem23796 | |
2-(trifluoromethyl)-1H-indole |
C9H6F3N | |
QFHVHZJGQWMBTE-UHFFFAOYSA-N | |
10932124 | |
C1=CC=C2C(=C1)C=C(N2)C(F)(F)F |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning