missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)phenol, TCI America™

Manufacturer:  TCI America T215625G

 View more versions of this product

Catalog No. T215625G



Specifications

Specifications

2-(Trifluoromethoxy)phenol
25g
C7H5F3O2
1993
GQWMNVOVQZIPJC-UHFFFAOYSA-N
2-(trifluoromethoxy)phenol
2777299
Clear Liquid at 20°C
Yellow
32858-93-8
MFCD00236324
2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluoromethoxy phenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
C1=CC=C(C(=C1)O)OC(F)(F)F
178.11
178.11
≥96.0% (GC)

Chemical Identifiers

32858-93-8
178.11
GQWMNVOVQZIPJC-UHFFFAOYSA-N
2777299
C1=CC=C(C(=C1)O)OC(F)(F)F
C7H5F3O2
MFCD00236324
2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluoromethoxy phenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
2-(trifluoromethoxy)phenol
Safety and Handling

Safety and Handling

TSCA : No

Documents