Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)phenol, 97%

$54.65 - $54.65

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethoxy)phenol
CAS 32858-93-8
Molecular Formula C7H5F3O2
Formula Weight 178.11
MDL Number MFCD00236324
Beilstein 1869013
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2037603

 
alfa aesar™
B2037603
1g Each for $54.65
Description & Specifications

Specifications

Formula Weight 178.11
Chemical Name or Material 2-(Trifluoromethoxy)phenol
Assay 97%
Molecular Formula C7H5F3O2
MDL Number MFCD00236324
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 69° to 71°C (60mmHg)
Quantity 1g
CAS 32858-93-8
Beilstein 1869013
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.