Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)-DL-phenylalanine, 97%

$78.04 - $265.19

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethoxy)-DL-phenylalanine
CAS 1218228-20-6
Molecular Formula C10H10F3NO3
Formula Weight 249.19
MDL Number MFCD07371965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3266703

  SDS
alfa aesar™
H3266703
1g Each for $265.19

AAH32667MD

  SDS
alfa aesar™
H32667MD
250mg Each for $78.04
Description & Specifications

Specifications

Formula Weight 249.19
Chemical Name or Material 2-(Trifluoromethoxy)-DL-phenylalanine
Assay 97%
Molecular Formula C10H10F3NO3
MDL Number MFCD07371965
CAS 1218228-20-6
Melting Point ≈236°C
Quantity 1g