missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)benzoyl Chloride, TCI America™

Manufacturer:  TCI America T23401G

 View more versions of this product

Catalog No. T23401G



Specifications

Specifications

2-(Trifluoromethoxy)benzoyl Chloride
Yellow
162046-61-9
MFCD00052330
2-trifluoromethoxy benzoyl chloride, o-trifluoromethoxybenzoyl chloride, trifluoromethoxybenzoyl chloride, 2-trifluoromethoxy benzoylchloride, 2-trifluoromethoxy-benzoyl chloride, benzoyl chloride, 2-trifluoromethoxy, 2-trifluoromethyloxy benzoyl chloride, pubchem10386, acmc-209dnm, ksc495e3j
C1=CC=C(C(=C1)C(=O)Cl)OC(F)(F)F
224.563
224.56
≥98.0% (GC)
96°C
1g
C8H4ClF3O2
3265
KSHPWYXAJJCUMJ-UHFFFAOYSA-N
2-(trifluoromethoxy)benzoyl chloride
2777241
Clear Liquid at 20°C

Chemical Identifiers

162046-61-9
224.563
KSHPWYXAJJCUMJ-UHFFFAOYSA-N
2777241
C1=CC=C(C(=C1)C(=O)Cl)OC(F)(F)F
C8H4ClF3O2
MFCD00052330
2-trifluoromethoxy benzoyl chloride, o-trifluoromethoxybenzoyl chloride, trifluoromethoxybenzoyl chloride, 2-trifluoromethoxy benzoylchloride, 2-trifluoromethoxy-benzoyl chloride, benzoyl chloride, 2-trifluoromethoxy, 2-trifluoromethyloxy benzoyl chloride, pubchem10386, acmc-209dnm, ksc495e3j
2-(trifluoromethoxy)benzoyl chloride
Safety and Handling

Safety and Handling

TSCA : No

Documents