missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ A1571003

 View more versions of this product

Catalog No. AAA1571003



Specifications

Specifications

2-(Trifluoromethoxy)benzonitrile
1g
C8H4F3NO
UN3276
ACNBBQGAWMHXLA-UHFFFAOYSA-N
2-(trifluoromethoxy)benzonitrile
2777232
97%
80°C to 82°C (15mmHg)
63968-85-4
MFCD00042409
2-trifluoromethoxy benzonitrile, o-trifluoromethoxybenzonitrile, benzonitrile, 2-trifluoromethoxy, 2-trifluoromethoxy benzenecarbonitrile, pubchem4824, acmc-1b9l5, ksc493e9p, o-trifluoromethoxy benzonitrile, acnbbqgawmhxla-uhfffaoysa, 2-trifluoromethoxy benzonitrile
C1=CC=C(C(=C1)C#N)OC(F)(F)F
187.121
187.12

Chemical Identifiers

63968-85-4
187.121
ACNBBQGAWMHXLA-UHFFFAOYSA-N
2777232
C1=CC=C(C(=C1)C#N)OC(F)(F)F
C8H4F3NO
MFCD00042409
2-trifluoromethoxy benzonitrile, o-trifluoromethoxybenzonitrile, benzonitrile, 2-trifluoromethoxy, 2-trifluoromethoxy benzenecarbonitrile, pubchem4824, acmc-1b9l5, ksc493e9p, o-trifluoromethoxy benzonitrile, acnbbqgawmhxla-uhfffaoysa, 2-trifluoromethoxy benzonitrile
2-(trifluoromethoxy)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Documents