Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)benzoic acid, 98%

$57.09 - $806.58

Chemical Identifiers

Chemical Name or Material 2- (Trifluoromethoxy)benzoic acid
CAS 1979-29-9
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD00052325
Beilstein 2645481
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1144403

  SDS
alfa aesar™
A1144403
1g Each for $57.09

AAA1144406

  SDS
alfa aesar™
A1144406
5g Each for $192.41

AAA1144414

  SDS
alfa aesar™
A1144414
25g Each for $806.58
Description & Specifications

Specifications

Formula Weight 206.12
Chemical Name or Material 2- (Trifluoromethoxy)benzoic acid
Assay 98%
Molecular Formula C8H5F3O3
MDL Number MFCD00052325
CAS 1979-29-9
Beilstein 2645481
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 78° to 80°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.