Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)benzoic acid, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL00372DA

 View more versions of this product

Catalog No. TL00372DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 206.12
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Linear Formula CF3OC6H4CO2H
Molecular Formula C8H5F3O3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-(Trifluoromethoxy)benzoic acid
Assay Percent Range 97%
MDL Number MFCD00052325
CAS 1979-29-9

Specifications

Chemical Name or Material 2-(Trifluoromethoxy)benzoic acid
CAS 1979-29-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H5F3O3
Linear Formula CF3OC6H4CO2H
Formula Weight 206.12
MDL Number MFCD00052325
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents