Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)benzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2- (Trifluoromethoxy)benzenesulfonyl chloride
CAS 103008-51-1
Molecular Formula C7H4ClF3O3S
Formula Weight 260.62
MDL Number MFCD00052316
UN Number UN3261
Beilstein 8482182
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1070703

  SDS
alfa aesar™
A1070703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1070706

  SDS
alfa aesar™
A1070706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 260.62
Chemical Name or Material 2- (Trifluoromethoxy)benzenesulfonyl chloride
Assay 97%
Molecular Formula C7H4ClF3O3S
MDL Number MFCD00052316
CAS 103008-51-1
UN Number UN3261
Beilstein 8482182
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 75° to 77°C (1mmHg)
Melting Point 29° to 32°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.