Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)benzeneboronic acid, 98%

$110.14 - $110.14

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethoxy)benzeneboronic acid
CAS 175676-65-0
Molecular Formula C7H6BF3O3
Formula Weight 205.93
MDL Number MFCD01320763
Beilstein 8766167
Synonym 2-(Trifluoromethoxy)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1977403

  SDS
alfa aesar™
L1977403
1g Each for $110.14
Description & Specifications

Specifications

Formula Weight 205.93
Chemical Name or Material 2-(Trifluoromethoxy)benzeneboronic acid
Synonym 2-(Trifluoromethoxy)phenylboronic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 118° to 120°C
Assay 98%
CAS 175676-65-0
Health Hazard 1 Warning
MDL Number MFCD01320763
Molecular Formula C7H6BF3O3
Quantity 1g
Beilstein 8766167
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.