Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethoxy)benzamide, 98%

$287.85 - $287.85

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethoxy)benzamide
CAS 127979-74-2
Molecular Formula C8H6F3NO2
Formula Weight 205.14
MDL Number MFCD00042407
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1388906

 
alfa aesar™
A1388906
5g Each for $287.85
Description & Specifications

Specifications

Formula Weight 205.14
Chemical Name or Material 2-(Trifluoromethoxy)benzamide
Assay 98%
Molecular Formula C8H6F3NO2
MDL Number MFCD00042407
CAS 127979-74-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 153° to 155°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.