missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)benzaldehyde, 96%, ACROS Organics™

Manufacturer:  Acros Organics AC433200050

 View more versions of this product

Catalog No. AC433200050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(Trifluoromethoxy)benzaldehyde
Colorless to Yellow
94651-33-9
100.0
C8H5F3O2
2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde
C1=CC=C(C(=C1)C=O)OC(F)(F)F
190.121
190.12
≥95.0%
Authentic
5g
95.0
96%
MFCD00042405
CPHXLFKIUVVIOQ-UHFFFAOYSA-N
2-(trifluoromethoxy)benzaldehyde
2777192
Liquid

Chemical Identifiers

94651-33-9
190.121
CPHXLFKIUVVIOQ-UHFFFAOYSA-N
2777192
C1=CC=C(C(=C1)C=O)OC(F)(F)F
C8H5F3O2
MFCD00042405
2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde
2-(trifluoromethoxy)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents