Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)benzaldehyde, 96%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC433200010

 View more versions of this product

Catalog No. AC433200010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥95.0%
Formula Weight 190.12
Quantity 1g
Color Clear colorless to pale yellow
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2-(Trifluoromethoxy)benzaldehyde
Assay Percent Range 96%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H5F3O2
MDL Number MFCD00042405
CAS 94651-33-9
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-(Trifluoromethoxy)benzaldehyde
CAS 94651-33-9
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD00042405
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents