missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)benzaldehyde, 96%, ACROS Organics™

Chemical Identifiers

CAS 94651-33-9
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD00042405
Chemical Name or Material 2-(Trifluoromethoxy)benzaldehyde
Assay ≥95.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC433200050 SDS Thermo Fisher Scientific
AC433200050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC433200010 SDS Thermo Fisher Scientific
AC433200010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

94651-33-9
100.0
C8H5F3O2
MFCD00042405
≥95.0%
95.0
96%
190.12
2-(Trifluoromethoxy)benzaldehyde

Specifications

190.12
5g
95.0
96%
MFCD00042405
Authentic
≥95.0%
Liquid
Clear colorless to pale yellow
100.0
94651-33-9
C8H5F3O2
2-(Trifluoromethoxy)benzaldehyde

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.