missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific Chemicals
$107.33 - $777.12
Chemical Identifiers
CAS | 94651-33-9 |
---|---|
Molecular Formula | C8H5F3O2 |
Molecular Weight (g/mol) | 190.12 |
MDL Number | MFCD00042405 |
InChI Key | CPHXLFKIUVVIOQ-UHFFFAOYSA-N |
Synonym | 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde |
PubChem CID | 2777192 |
IUPAC Name | 2-(trifluoromethoxy)benzaldehyde |
SMILES | FC(F)(F)OC1=CC=CC=C1C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2084006
|
Thermo Scientific Chemicals
B2084006 |
5 g |
Each for $107.33
|
|
|||||
AAB2084014
|
Thermo Scientific Chemicals
B2084014 |
25 g |
Each for $354.48
|
|
|||||
AAB2084022
|
Thermo Scientific Chemicals
B2084022 |
100 g |
Each for $777.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
94651-33-9 | |
190.12 | |
CPHXLFKIUVVIOQ-UHFFFAOYSA-N | |
2777192 | |
FC(F)(F)OC1=CC=CC=C1C=O |
C8H5F3O2 | |
MFCD00042405 | |
2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde | |
2-(trifluoromethoxy)benzaldehyde |
Specifications
94651-33-9 | |
77°C (20 mmHg) | |
C8H5F3O2 | |
MFCD00042405 | |
6137162 | |
2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde | |
FC(F)(F)OC1=CC=CC=C1C=O | |
190.12 | |
190.12 | |
2-(Trifluoromethoxy)benzaldehyde |
1.33 | |
67°C (152°F) | |
1.455 | |
5 g | |
Air Sensitive | |
CPHXLFKIUVVIOQ-UHFFFAOYSA-N | |
2-(trifluoromethoxy)benzaldehyde | |
2777192 | |
96% |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only