Please login to your online account to display your discounted pricing

2-(Trifluoromethoxy)aniline, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  297620050

 View more versions of this product

Catalog No. AC297620050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 177.12
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
EINECS Number 216-257-9
TSCA TSCA
CAS Max % 100.0
Refractive Index 1.4605 to 1.4625 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Chemical Name or Material 2-(Trifluoromethoxy)aniline
Linear Formula CF3OC6H4NH2
Molecular Formula C7H6F3NO
CAS 1535-75-7
MDL Number MFCD00035959
Assay Percent Range 99%

Specifications

Chemical Name or Material 2-(Trifluoromethoxy)aniline
Name Note 99%
CAS 1535-75-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6F3NO
Linear Formula CF3OC6H4NH2
Formula Weight 177.12
MDL Number MFCD00035959
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents