missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-(Trifluoromethoxy)acetophenone 98.0+%, TCI America™
Supplier: TCI America T279425G
Specifications
2′-(Trifluoromethoxy)acetophenone | |
Yellow | |
MFCD00236320 | |
2'-trifluoromethoxy acetophenone, 1-2-trifluoromethoxy phenyl ethanone, 1-2-trifluoromethoxy phenyl ethan-1-one, 1-acetyl-2-trifluoromethoxy benzene, pubchem4295, 2-trifluoromethoxy acetophenone98, 1-2-trifluoromethoxyphenyl ethanone, 2'-trifluoromethoxy acetophenone98 | |
CC(=O)C1=CC=CC=C1OC(F)(F)F | |
204.148 | |
204.15 | |
Liquid |
220227-93-0 | |
C9H7F3O2 | |
25 g | |
XTXSTESGCXKUIH-UHFFFAOYSA-N | |
1-[2-(trifluoromethoxy)phenyl]ethanone | |
2777176 | |
≥98.0% (GC) |
Chemical Identifiers
220227-93-0 | |
204.148 | |
XTXSTESGCXKUIH-UHFFFAOYSA-N | |
2777176 | |
CC(=O)C1=CC=CC=C1OC(F)(F)F |
C9H7F3O2 | |
MFCD00236320 | |
2'-trifluoromethoxy acetophenone, 1-2-trifluoromethoxy phenyl ethanone, 1-2-trifluoromethoxy phenyl ethan-1-one, 1-acetyl-2-trifluoromethoxy benzene, pubchem4295, 2-trifluoromethoxy acetophenone98, 1-2-trifluoromethoxyphenyl ethanone, 2'-trifluoromethoxy acetophenone98 | |
1-[2-(trifluoromethoxy)phenyl]ethanone |
Safety and Handling
TSCA : No