Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-(Trifluoromethoxy)acetophenone, 98+%

$62.34 - $223.19

Chemical Identifiers

Chemical Name or Material 2'-(Trifluoromethoxy)acetophenone
CAS 220227-93-0
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD00236320
Beilstein 2259035
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1990603

 
alfa aesar™
L1990603
1g Each for $62.34

AAL1990606

 
alfa aesar™
L1990606
5g Each for $223.19
Description & Specifications

Specifications

Formula Weight 204.15
Chemical Name or Material 2'-(Trifluoromethoxy)acetophenone
Assay ≥98%
Molecular Formula C9H7F3O2
MDL Number MFCD00236320
CAS 220227-93-0
Beilstein 2259035
Quantity 1g