missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoroacetyl)thiophene, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC434700010
Specifications
2-(Trifluoroacetyl)thiophene | |
Authentic | |
163.0°C to 164.0°C | |
1g | |
97.5 | |
97.5% min. (GC) | |
2,2,2-trifluoro-1-2-thienyl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethanone, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, 2-trifluoroacetyl thiophene, 2-trifluoroacetylthiophene, acmc-20aolw, ethanone, 2,2,2-trifluoro-1-2-thienyl, ethanone,2,2,2-trifluoro-1-2-thienyl, trifluoroacetylthiophene | |
C1=CSC(=C1)C(=O)C(F)(F)F | |
180.15 | |
180.15 | |
98% |
59°C | |
Glass Bottle | |
Colorless to Yellow | |
651-70-7 | |
100.0 | |
C6H3F3OS | |
CZYKJGCKVBXLGF-UHFFFAOYSA-N | |
2,2,2-trifluoro-1-thiophen-2-ylethanone | |
136464 | |
Liquid |
Chemical Identifiers
651-70-7 | |
180.15 | |
2,2,2-trifluoro-1-2-thienyl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethanone, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, 2-trifluoroacetyl thiophene, 2-trifluoroacetylthiophene, acmc-20aolw, ethanone, 2,2,2-trifluoro-1-2-thienyl, ethanone,2,2,2-trifluoro-1-2-thienyl, trifluoroacetylthiophene | |
2,2,2-trifluoro-1-thiophen-2-ylethanone |
C6H3F3OS | |
CZYKJGCKVBXLGF-UHFFFAOYSA-N | |
136464 | |
C1=CSC(=C1)C(=O)C(F)(F)F |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning