missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%

Manufacturer:  Alfa Aesar™ L1358706

 View more versions of this product

Catalog No. AAL1358706



Specifications

Specifications

2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride
5g
C11H9ClF3NO3S
UN3261
2-2,2,2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-2,2,2-trifluoroacetyl-3,4-dihydro-1h-isoquinoline-7-sulfonyl chloride, 1-7-chlorosulfonyl 2-1,2,3,4-tetrahydroisoquinolyl-2,2,2-trifluoroethan-1-one, 2-trifluoroacetyl-7-chlorosulfonyl-1,2,3,4-tetrahydroisoquinoline, 7-chlorosulfonyl-2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfone
C1CN(CC2=C1C=CC(=C2)S(=O)(=O)Cl)C(=O)C(F)(F)F
327.702
327.71
102°C to 106°C
74291-57-9
MFCD01075651
5603124
SXIYEBVAQTUBOQ-UHFFFAOYSA-N
2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride
11324902
99%

Chemical Identifiers

74291-57-9
327.702
SXIYEBVAQTUBOQ-UHFFFAOYSA-N
11324902
C1CN(CC2=C1C=CC(=C2)S(=O)(=O)Cl)C(=O)C(F)(F)F
C11H9ClF3NO3S
MFCD01075651
2-2,2,2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-2,2,2-trifluoroacetyl-3,4-dihydro-1h-isoquinoline-7-sulfonyl chloride, 1-7-chlorosulfonyl 2-1,2,3,4-tetrahydroisoquinolyl-2,2,2-trifluoroethan-1-one, 2-trifluoroacetyl-7-chlorosulfonyl-1,2,3,4-tetrahydroisoquinoline, 7-chlorosulfonyl-2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfone
2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents