Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Tridecanone, 98+%

$25.32 - $139.68

Chemical Identifiers

Chemical Name or Material 2-Tridecanone
CAS 593-08-8
Molecular Formula C13H26O
Formula Weight 198.35
MDL Number MFCD00008968
UN Number UN3082
Beilstein 1757402
Synonym Methyl n-undecyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1904214

  SDS
alfa aesar™
A1904214
25g Each for $25.32

AAA1904222

  SDS
alfa aesar™
A1904222
100g Each for $139.68
Description & Specifications

Specifications

Formula Weight 198.35
Chemical Name or Material 2-Tridecanone
TSCA TSCA
Assay ≥98%
Molecular Formula C13H26O
MDL Number MFCD00008968
CAS 593-08-8
UN Number UN3082
Beilstein 1757402
Synonym Methyl n-undecyl ketone
Odor Characteristic
Boiling Point 133° to 134°C (10mmHg)
Melting Point 29° to 31°C
Flash Point 122°C (251°F)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Quantity 25g