Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Tridecanol, 97%

$33.76 - $96.58

Chemical Identifiers

Chemical Name or Material 2-Tridecanol
CAS 1653-31-2
Molecular Formula C13H28O
Formula Weight 200.37
MDL Number MFCD00037601
Beilstein 1720217
Synonym Methyl undecyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0192603

  SDS
alfa aesar™
L0192603
1g Each for $33.76

AAL0192606

  SDS
alfa aesar™
L0192606
5g Each for $96.58
Description & Specifications

Specifications

Formula Weight 200.37
Chemical Name or Material 2-Tridecanol
Assay 97%
Molecular Formula C13H28O
MDL Number MFCD00037601
CAS 1653-31-2
Beilstein 1720217
Synonym Methyl undecyl carbinol
Boiling Point 265°C
Melting Point 23° to 26°C
Quantity 1g