missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Tri-n-butylstannyl)thiophene, 97%

Manufacturer:  Alfa Aesar™ H55904AA

 View more versions of this product

Catalog No. AAH55904AA



Description

Description

Applications
2-(Tri-n-butylstannyl)thiophene derivative is employed in Stille reaction involving efficient carbon-carbon bond formation.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2-(Tri-n-butylstannyl)thiophene
1.518
Colorless to Yellow
54663-78-4
MFCD00192513
2-thienyltributyltin
CCCC[Sn](CCCC)(CCCC)C1=CC=CS1
373.186
373.16
97%
1.175 g/mL
155°C (0.1mmHg)
5mL
C16H30SSn
UN2788
UKTDFYOZPFNQOQ-UHFFFAOYSA-N
tributyl(thiophen-2-yl)stannane
2779373
Liquid

Chemical Identifiers

54663-78-4
373.186
UKTDFYOZPFNQOQ-UHFFFAOYSA-N
2779373
CCCC[Sn](CCCC)(CCCC)C1=CC=CS1
C16H30SSn
MFCD00192513
2-thienyltributyltin
tributyl(thiophen-2-yl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents