Learn More
2-(Tri-n-butylstannyl)thiazole, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5109003
Description
Used as reagent for arylation of thiazole by Stille cross-coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as reagent for arylation of thiazole by Stille cross-coupling.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
2-(Tri-n-butylstannyl)thiazole | |
1.19 | |
C15H29NSSn | |
1 g | |
2-tributylstannyl thiazole, 2-tributylstannylthiazole, 2-tributylstannyl-1,3-thiazole, thiazole,2-tributylstannyl, tributylstannylthiazole, thiazole, 2-tributylstannyl, 2-tributylstannanyl-thiazole, 1,3-thiazol-2-yl tributylstannane, 2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl thiazole | |
WUOFQGMXQCSPPV-UHFFFAOYSA-N | |
tributyl(1,3-thiazol-2-yl)stannane | |
2763259 | |
96% |
121359-48-6 | |
307°C to 309°C | |
MFCD01319057 | |
UN2788 | |
Not miscible or difficult to mix. | |
CCCC[Sn](CCCC)(CCCC)C1=NC=CS1 | |
374.174 | |
374.17 |
Chemical Identifiers
121359-48-6 | |
374.174 | |
WUOFQGMXQCSPPV-UHFFFAOYSA-N | |
2763259 | |
CCCC[Sn](CCCC)(CCCC)C1=NC=CS1 |
C15H29NSSn | |
MFCD01319057 | |
2-tributylstannyl thiazole, 2-tributylstannylthiazole, 2-tributylstannyl-1,3-thiazole, thiazole,2-tributylstannyl, tributylstannylthiazole, thiazole, 2-tributylstannyl, 2-tributylstannanyl-thiazole, 1,3-thiazol-2-yl tributylstannane, 2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl thiazole | |
tributyl(1,3-thiazol-2-yl)stannane |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only